Product Details

SNP ID
rs9944968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9549662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGTGTGCACAGTATGTGTGTGG[G/T]GGGGTACATGTATGTATGCCTGTGT
Phenotype
MIM: 604908
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP4R1 PubMed Links

Gene Details

Gene
PPP4R1
Gene Name
protein phosphatase 4 regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042388.2 Intron NP_001035847.1
NM_005134.3 Intron NP_005125.1
XM_011525775.2 Intron XP_011524077.1
XM_011525776.1 Intron XP_011524078.1
XM_011525779.2 Intron XP_011524081.1
XM_017026105.1 Intron XP_016881594.1
XM_017026106.1 Intron XP_016881595.1
XM_017026107.1 Intron XP_016881596.1

View Full Product Details