Product Details
- SNP ID
-
rs6458307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42763377 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCTTTTATATAAATGCTCATTCA[C/T]TGAGTTCAAATACTTTTAAAATGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLTSCR1L
PubMed Links
Gene Details
- Gene
- GLTSCR1L
- Gene Name
- GLTSCR1 like
View Full Product Details