Product Details

SNP ID
rs9880206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172444137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTCCTGAATATTAACACATG[C/G]AACTCAACTTAATTTAAAAATGTAT
Phenotype
MIM: 601898
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GHSR PubMed Links

Gene Details

Gene
GHSR
Gene Name
growth hormone secretagogue receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004122.2 2168 Intron NP_004113.1
NM_198407.2 2168 UTR 3 NP_940799.1

View Full Product Details