Product Details

SNP ID
rs9859712
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169085490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGACTCCTACACTTTATAAC[C/G]GAAAGCACTGAAGGTGCTTGAGTGA
Phenotype
MIM: 165215
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MECOM PubMed Links

Gene Details

Gene
MECOM
Gene Name
MDS1 and EVI1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105077.3 Intron NP_001098547.3
NM_001105078.3 Intron NP_001098548.2
NM_001163999.1 Intron NP_001157471.1
NM_001164000.1 Intron NP_001157472.1
NM_001205194.1 Intron NP_001192123.1
NM_004991.3 Intron NP_004982.2
NM_005241.3 Intron NP_005232.2
XM_005247213.3 Intron XP_005247270.1
XM_005247214.3 Intron XP_005247271.1
XM_005247215.3 Intron XP_005247272.1
XM_005247219.2 Intron XP_005247276.1
XM_005247220.2 Intron XP_005247277.1
XM_005247221.2 Intron XP_005247278.1
XM_005247223.2 Intron XP_005247280.1
XM_005247224.3 Intron XP_005247281.1
XM_005247225.3 Intron XP_005247282.1
XM_005247226.3 Intron XP_005247283.1
XM_011512546.2 Intron XP_011510848.1
XM_017005874.1 Intron XP_016861363.1
XM_017005875.1 Intron XP_016861364.1
XM_017005876.1 Intron XP_016861365.1
XM_017005877.1 Intron XP_016861366.1
XM_017005878.1 Intron XP_016861367.1

View Full Product Details