Product Details
- SNP ID
-
rs4242182
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:174729165 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTCTCTCTGTTCTCTAGGACATA[C/T]GAGCCCTACCACCTGCACCCTGAGG
- Phenotype
-
MIM: 123101
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MSX2
PubMed Links
Gene Details
- Gene
- MSX2
- Gene Name
- msh homeobox 2
View Full Product Details