Product Details

SNP ID
rs7200817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4430597 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAGAAAGCAGTTGTTGGACA[C/G]TGGGTTGGCCATTCTTAGAACGACT
Phenotype
MIM: 608382
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA3 PubMed Links

Gene Details

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135110.2 Intron NP_001128582.1
NM_001286516.1 Intron NP_001273445.1
NM_005147.5 Intron NP_005138.3

View Full Product Details