Product Details
- SNP ID
-
rs9918716
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:4658320 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAAGCTCCCCTCCCAGAGTTTGA[A/T]TCAGTGGATCAGGGGTGGGACCTGA
- Phenotype
-
MIM: 616302
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXK1
PubMed Links
Gene Details
- Gene
- FOXK1
- Gene Name
- forkhead box K1
View Full Product Details