Product Details

SNP ID
rs10203154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200490375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGATGCCCAGGTGCCCCCGTGC[A/C]CACCAGGGCCGTGGCTCTGGAAGGT
Phenotype
MIM: 613817
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCTD18 PubMed Links

Gene Details

Gene
KCTD18
Gene Name
potassium channel tetramerization domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321547.1 1548 Missense Mutation GGC,TGC G336C NP_001308476.1
NM_001321548.1 1548 Missense Mutation GGC,TGC G127C NP_001308477.1
NM_001321550.1 1548 Missense Mutation GGC,TGC G127C NP_001308479.1
NM_152387.3 1548 Missense Mutation GGC,TGC G336C NP_689600.2
Gene
SPATS2L
Gene Name
spermatogenesis associated serine rich 2 like
There are no transcripts associated with this gene.

View Full Product Details