Product Details

SNP ID
rs10105305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39937236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGAATGATGGCTCCCAGTTTATC[A/T]TGCAGGTGGGATTATATTCCAACAG
Phenotype
MIM: 147435 MIM: 612129
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IDO1 PubMed Links

Gene Details

Gene
IDO1
Gene Name
indoleamine 2,3-dioxygenase 1
There are no transcripts associated with this gene.

Gene
IDO2
Gene Name
indoleamine 2,3-dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194294.2 Intron NP_919270.2

View Full Product Details