Product Details

SNP ID
rs10142126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:65426240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCTTTTTCTGCCAGTCCTATTT[A/T]ACTTAGCCTAATGTCCTTCGGGTTC
Phenotype
MIM: 602589
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FUT8 PubMed Links

Gene Details

Gene
FUT8
Gene Name
fucosyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004480.4 Intron NP_004471.4
NM_178155.2 Intron NP_835368.1
NM_178156.2 Intron NP_835369.1
XM_011536614.2 Intron XP_011534916.1
XM_017021136.1 Intron XP_016876625.1
XM_017021137.1 Intron XP_016876626.1
XM_017021138.1 Intron XP_016876627.1
XM_017021139.1 Intron XP_016876628.1
XM_017021140.1 Intron XP_016876629.1

View Full Product Details