Product Details

SNP ID
rs9607505
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37972367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTAATGCAGAGTTAATAGGGG[C/T]TAGAGTGGCTAGGAGAGGGGACTAC
Phenotype
MIM: 604414 MIM: 602229
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR6820 PubMed Links

Gene Details

Gene
MIR6820
Gene Name
microRNA 6820
There are no transcripts associated with this gene.

Gene
POLR2F
Gene Name
RNA polymerase II subunit F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301129.1 2807 Intron NP_001288058.1
NM_001301130.1 2807 Intron NP_001288059.1
NM_001301131.1 2807 Intron NP_001288060.1
NM_021974.4 2807 Intron NP_068809.1
XM_017028832.1 2807 Intron XP_016884321.1
Gene
SOX10
Gene Name
SRY-box 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006941.3 2807 UTR 3 NP_008872.1

View Full Product Details