Product Details
- SNP ID
-
rs7200854
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:72049419 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTTCTAACAGATGAAAAGGTTAG[A/G]CCAAAAAATTCTATATAAAGAATGA
- Phenotype
-
MIM: 140100
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HP
PubMed Links
Gene Details
- Gene
- HP
- Gene Name
- haptoglobin
There are no transcripts associated with this gene.
- Gene
- TXNL4B
- Gene Name
- thioredoxin like 4B
View Full Product Details