Product Details

SNP ID
rs5918177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41450594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGATTTGCATTTTAAAGGAAGT[C/T]TATAGGAAAATAAAATCACAATGAG
Phenotype
MIM: 300278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NYX PubMed Links

Gene Details

Gene
NYX
Gene Name
nyctalopin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022567.2 Intron NP_072089.1

View Full Product Details