Product Details

SNP ID
rs6031620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44480765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTGCCTGACGGTCTGTGTATCC[C/T]TCAGCCACACATTTCTGCCTTTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTPAL PubMed Links

Gene Details

Gene
TTPAL
Gene Name
alpha tocopherol transfer protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039199.2 Intron NP_001034288.1
NM_001261839.1 Intron NP_001248768.1
NM_024331.4 Intron NP_077307.2
XM_011529043.2 Intron XP_011527345.1
XM_011529044.2 Intron XP_011527346.1
XM_011529045.2 Intron XP_011527347.1
XM_017028063.1 Intron XP_016883552.1

View Full Product Details