Product Details

SNP ID
rs9791154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149498373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATATACTTCACCATAGGGAATA[C/T]CATTTGATGCATGTTATTAAGAAGT
Phenotype
MIM: 600505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSNK1A1 PubMed Links

Gene Details

Gene
CSNK1A1
Gene Name
casein kinase 1 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025105.2 Intron NP_001020276.1
NM_001271741.1 Intron NP_001258670.1
NM_001271742.1 Intron NP_001258671.1
NM_001892.5 Intron NP_001883.4

View Full Product Details