Product Details

SNP ID
rs10149100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91781971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGTAAGAATGGGGGCAGAAAAA[A/C]AGGGAAGACTGAGAACACAGAGCAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TC2N PubMed Links

Gene Details

Gene
TC2N
Gene Name
tandem C2 domains, nuclear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128595.2 2814 UTR 3 NP_001122067.1
NM_001128596.2 2814 UTR 3 NP_001122068.1
NM_001289134.1 2814 UTR 3 NP_001276063.1
NM_152332.5 2814 UTR 3 NP_689545.1

View Full Product Details