Product Details
- SNP ID
-
rs5978102
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135169937 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAATCCACAACAATCACAATTTCG[C/T]CTAATAGGGTTGTTGTAGGTTTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CT55
PubMed Links
Gene Details
- Gene
- CT55
- Gene Name
- cancer/testis antigen 55
View Full Product Details