Product Details

SNP ID
rs9905415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46767244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGCTTGGGAACAAAATCCACC[C/T]GCCTTTGATCCCTCGATTACTCATC
Phenotype
MIM: 601633 MIM: 165330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSF PubMed Links

Gene Details

Gene
NSF
Gene Name
N-ethylmaleimide sensitive factor, vesicle fusing ATPase
There are no transcripts associated with this gene.

Gene
WNT3
Gene Name
Wnt family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030753.4 Intron NP_110380.1

View Full Product Details