Product Details

SNP ID
rs10171382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46301148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCATTTCAAGTTTTTTTCAGTGG[G/T]GCTTATAGGGAGCCTAGAATTATTT
Phenotype
MIM: 603349
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EPAS1 PubMed Links

Gene Details

Gene
EPAS1
Gene Name
endothelial PAS domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001430.4 Intron NP_001421.2
XM_011532698.2 Intron XP_011531000.1

View Full Product Details