Product Details

SNP ID
rs10498187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:225406089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTCTAACTCAATACTCACAACA[A/T]GGGTCTATTCCAACCACTATTTTTT
Phenotype
MIM: 615478
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NYAP2 PubMed Links

Gene Details

Gene
NYAP2
Gene Name
neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020864.1 Intron NP_065915.1
XM_005246708.3 Intron XP_005246765.1
XM_011511524.1 Intron XP_011509826.1
XM_017004564.1 Intron XP_016860053.1

View Full Product Details