Product Details

SNP ID
rs5993467
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19023197 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTAAGGGTCGAGGTGCAGCCACA[A/G]CCTGGCTCTGTGCACACCCCCTCCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DGCR10 PubMed Links
Additional Information
For this assay, SNP(s) [rs116558919] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DGCR10
Gene Name
DiGeorge syndrome critical region gene 10 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR5
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR9
Gene Name
DiGeorge syndrome critical region gene 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details