Product Details
- SNP ID
-
rs10012401
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:168095377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAATCCTATTCCCTTTTCTACCC[C/T]TTTGACACCTTTTCCTACTTCTATA
- Phenotype
-
MIM: 608008
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA10
PubMed Links
Gene Details
- Gene
- ANXA10
- Gene Name
- annexin A10
View Full Product Details