Product Details
- SNP ID
-
rs10187661
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:106073832 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATGTATAACAGGGATTCACCGCAA[C/G]TGAGGAAGGAAGTCATTCTTTGTGC
- Phenotype
-
MIM: 611752
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72034828] are located under a probe and SNP(s) [rs137908864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf40
- Gene Name
- chromosome 2 open reading frame 40
View Full Product Details