Product Details

SNP ID
rs4142417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:7978861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCAGATCTAGCAAGAGACAAATA[A/G]TAAGAGAAACCAACATAAAGGTAAT
Phenotype
MIM: 616766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMX4 PubMed Links

Gene Details

Gene
TMX4
Gene Name
thioredoxin related transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021156.3 4656 UTR 3 NP_066979.2

View Full Product Details