Product Details
- SNP ID
-
rs6065928
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:37299008 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTTGGCCTGTCAAAGTGTTGGG[C/T]TTACACTTTGGGCGTAACCCACCAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MANBAL
PubMed Links
Gene Details
- Gene
- MANBAL
- Gene Name
- mannosidase beta like
View Full Product Details