Product Details

SNP ID
rs5757243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38686038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTGGCCCCAAGGTCCTGCTTG[A/T]GACAATCAGCAAGGTTGGCTGTAAT
Phenotype
MIM: 615323 MIM: 607046
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JOSD1 PubMed Links

Gene Details

Gene
JOSD1
Gene Name
Josephin domain containing 1
There are no transcripts associated with this gene.

Gene
TOMM22
Gene Name
translocase of outer mitochondrial membrane 22
There are no transcripts associated with this gene.

View Full Product Details