Product Details
- SNP ID
-
rs10146397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55578851 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCAATATCTCAGGCCACTATACA[C/T]TGCGCAGTTTTCATTTGTATTTTGG
- Phenotype
-
MIM: 600381
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KTN1
PubMed Links
Gene Details
- Gene
- KTN1
- Gene Name
- kinectin 1
There are no transcripts associated with this gene.
- Gene
- KTN1-AS1
- Gene Name
- KTN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details