Product Details

SNP ID
rs9809001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50090861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACAGTCTGGCACTTAGCACAGT[A/T]CCTGGCTGTGGTAAGCAATCAATGA
Phenotype
MIM: 606884
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBM5 PubMed Links

Gene Details

Gene
RBM5
Gene Name
RNA binding motif protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005778.3 Intron NP_005769.1
XM_006712917.1 Intron XP_006712980.1
XM_011533261.1 Intron XP_011531563.1
XM_011533262.1 Intron XP_011531564.1
XM_017005503.1 Intron XP_016860992.1
XM_017005504.1 Intron XP_016860993.1
XM_017005505.1 Intron XP_016860994.1
XM_017005506.1 Intron XP_016860995.1
XM_017005507.1 Intron XP_016860996.1
Gene
RBM5-AS1
Gene Name
RBM5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details