Product Details

SNP ID
rs10419275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53268708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATATTATATATACTTTGTAAA[A/C]GGAAATTAGAGAGTTTGGGTGATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VN1R2 PubMed Links

Gene Details

Gene
VN1R2
Gene Name
vomeronasal 1 receptor 2
There are no transcripts associated with this gene.

Gene
VN1R4
Gene Name
vomeronasal 1 receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173857.2 Intron NP_776256.2
XM_011526885.1 Intron XP_011525187.1
XM_017026728.1 Intron XP_016882217.1
XM_017026729.1 Intron XP_016882218.1
XM_017026730.1 Intron XP_016882219.1

View Full Product Details