Product Details

SNP ID
rs9273652
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32661426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGAAGCCTCCAACGCCACTCAG[A/C]ATCTTGCTCTGGGCAGATTCAGACT
Phenotype
MIM: 146880 MIM: 604305
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs9273651] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
There are no transcripts associated with this gene.

Gene
HLA-DQB1
Gene Name
major histocompatibility complex, class II, DQ beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243961.1 775 Missense Mutation ATG,ATT M231I NP_001230890.1
NM_001243962.1 775 Missense Mutation ATG,ATT M231I NP_001230891.1
NM_002123.4 775 Missense Mutation ATG,ATT M231I NP_002114.3
Gene
HLA-DQB1-AS1
Gene Name
HLA-DQB1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details