Product Details

SNP ID
rs10400842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100883006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAACAGTCTGATTTTTTTTTAA[A/G]GTTGCTGGTTACAGATCAATGTCAT
Phenotype
MIM: 600463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH1A3 PubMed Links

Gene Details

Gene
ALDH1A3
Gene Name
aldehyde dehydrogenase 1 family member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000693.3 Intron NP_000684.2
NM_001293815.1 Intron NP_001280744.1
Gene
LOC101927751
Gene Name
uncharacterized LOC101927751
There are no transcripts associated with this gene.

Gene
LOC105369201
Gene Name
uncharacterized LOC105369201
There are no transcripts associated with this gene.

View Full Product Details