Product Details

SNP ID
rs9662723
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:145689573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAGCTTTTTCTTTTATAGGAGA[G/A]TAAGAACAGGGGCCAGAACAGCAAG
Phenotype
MIM: 603831
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PDZK1 PubMed Links

Gene Details

Gene
PDZK1
Gene Name
PDZ domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201325.1 Intron NP_001188254.1
NM_001201326.1 Intron NP_001188255.1
NM_002614.4 Intron NP_002605.2
XM_011509616.2 Intron XP_011507918.1
XM_011509618.2 Intron XP_011507920.1
XM_011509619.2 Intron XP_011507921.2
XM_017001464.1 Intron XP_016856953.1
XM_017001465.1 Intron XP_016856954.1
XM_017001466.1 Intron XP_016856955.1
XM_017001467.1 Intron XP_016856956.1

View Full Product Details