Product Details

SNP ID
rs9900392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49602463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGGTGAAAGAATTGGTGTCT[A/C]CCCTGTGAAATCCCTTATTTACTGC
Phenotype
MIM: 602650
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPOP PubMed Links

Gene Details

Gene
SPOP
Gene Name
speckle type BTB/POZ protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007226.1 Intron NP_001007227.1
NM_001007227.1 Intron NP_001007228.1
NM_001007228.1 Intron NP_001007229.1
NM_001007229.1 Intron NP_001007230.1
NM_001007230.1 Intron NP_001007231.1
NM_003563.3 Intron NP_003554.1
XM_005257723.4 Intron XP_005257780.1
XM_005257724.4 Intron XP_005257781.1
XM_017025204.1 Intron XP_016880693.1

View Full Product Details