Product Details

SNP ID
rs10249081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124828465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGAACCCATATTTAAACAGTA[G/T]AACTAATTAACAGGGAAAGTAAGAA
Phenotype
MIM: 606478
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POT1 PubMed Links

Gene Details

Gene
POT1
Gene Name
protection of telomeres 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042594.1 Intron NP_001036059.1
NM_015450.2 Intron NP_056265.2
XM_006715917.3 Intron XP_006715980.1
XM_011516006.1 Intron XP_011514308.1
XM_011516007.1 Intron XP_011514309.1
XM_017011941.1 Intron XP_016867430.1
XM_017011942.1 Intron XP_016867431.1

View Full Product Details