Product Details

SNP ID
rs9449441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82172760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCAGTGACCTACGATGAGCGAA[C/T]CTGGAAGCTTTCGCAGAGATTACAG
Phenotype
MIM: 606457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IBTK PubMed Links

Gene Details

Gene
IBTK
Gene Name
inhibitor of Bruton tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300906.1 Intron NP_001287835.1
NM_015525.3 Intron NP_056340.2
XM_006715453.2 Intron XP_006715516.1
XM_006715454.2 Intron XP_006715517.1

View Full Product Details