Product Details

SNP ID
rs5935654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13699464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGACAAGTGAATGGATGCAATTA[C/G]GTGTTGTGTGTCCCTCCCAGGGAGT
Phenotype
MIM: 300284
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB9A PubMed Links

Gene Details

Gene
RAB9A
Gene Name
RAB9A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195328.1 Intron NP_001182257.1
NM_004251.4 Intron NP_004242.1

View Full Product Details