Product Details
- SNP ID
-
rs6557412
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:155231692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCCTGACCTGGCTGCAGTGGGGT[A/G]GCCCAGGGTAGCTGCAGCTGGATGT
- Phenotype
-
MIM: 607033
MIM: 604709
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TFB1M
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143251421] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFB1M
- Gene Name
- transcription factor B1, mitochondrial
- Gene
- TIAM2
- Gene Name
- T-cell lymphoma invasion and metastasis 2
View Full Product Details