Product Details
- SNP ID
-
rs9536008
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:52315389 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACTTTTCAACCTGTAAACTGCTG[G/T]ATCCTACGGAATTCACATGTGTATT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TPTE2P2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139704985] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TPTE2P2
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details