Product Details

SNP ID
rs9903350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78186242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATAGCAAGATCCTACCTCTAGAA[G/T]AACAAAAACAAAACAAAAAGAATGG
Phenotype
MIM: 188300
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AFMID PubMed Links

Gene Details

Gene
AFMID
Gene Name
arylformamidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010982.4 Intron NP_001010982.2
NM_001145526.2 Intron NP_001138998.1
XM_011524326.2 Intron XP_011522628.1
XM_011524327.2 Intron XP_011522629.1
XM_011524328.2 Intron XP_011522630.1
XM_011524329.2 Intron XP_011522631.1
XM_011524330.2 Intron XP_011522632.1
XM_011524331.2 Intron XP_011522633.1
XM_011524332.2 Intron XP_011522634.1
XM_011524333.2 Intron XP_011522635.1
XM_011524334.2 Intron XP_011522636.1
XM_011524335.2 Intron XP_011522637.1
XM_011524336.2 Intron XP_011522638.1
XM_011524337.2 Intron XP_011522639.1
Gene
TK1
Gene Name
thymidine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003258.4 Intron NP_003249.3
XM_017024992.1 Intron XP_016880481.1

View Full Product Details