Product Details

SNP ID
rs9894979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29614787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTTTGCTACCTAACTTTACTTG[G/T]GGGGAACGCCTACGGTGTGGGTTGG
Phenotype
MIM: 615124
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD13B PubMed Links

Gene Details

Gene
ANKRD13B
Gene Name
ankyrin repeat domain 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152345.4 2306 Intron NP_689558.4
XM_005257920.4 2306 Intron XP_005257977.1
XM_011524319.2 2306 Intron XP_011522621.1
XM_011524320.2 2306 Intron XP_011522622.1
XM_011524321.2 2306 Intron XP_011522623.1
XM_017024174.1 2306 Intron XP_016879663.1
XM_017024175.1 2306 Intron XP_016879664.1
XM_017024176.1 2306 Intron XP_016879665.1
Gene
CORO6
Gene Name
coronin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032854.3 2306 UTR 3 NP_116243.2
XM_005258047.3 2306 UTR 3 XP_005258104.1
XM_005258048.4 2306 UTR 3 XP_005258105.2
XM_005258056.4 2306 UTR 3 XP_005258113.1
XM_011525385.2 2306 UTR 3 XP_011523687.1
XM_011525386.2 2306 UTR 3 XP_011523688.1
XM_011525387.2 2306 UTR 3 XP_011523689.1
XM_011525388.2 2306 Intron XP_011523690.1
XM_011525389.2 2306 Intron XP_011523691.1
XM_011525390.2 2306 UTR 3 XP_011523692.1
XM_011525391.2 2306 UTR 3 XP_011523693.1
XM_017025235.1 2306 UTR 3 XP_016880724.1

View Full Product Details