Product Details

SNP ID
rs6053077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5073924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAAGCTCTACAGGAAGCATGGCA[C/T]CAGCCAGGGCTCAGCTTCTGGTGAG
Phenotype
MIM: 617019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM230 PubMed Links

Gene Details

Gene
TMEM230
Gene Name
transmembrane protein 230
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009923.1 Intron NP_001009923.1
NM_001009924.1 Intron NP_001009924.1
NM_001009925.1 Intron NP_001009925.1
NM_014145.4 Intron NP_054864.3
XM_005260713.3 Intron XP_005260770.1
XM_011529227.2 Intron XP_011527529.1
XM_011529229.2 Intron XP_011527531.1
XM_017027834.1 Intron XP_016883323.1
XM_017027835.1 Intron XP_016883324.1
XM_017027836.1 Intron XP_016883325.1

View Full Product Details