Product Details
- SNP ID
-
rs5756589
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37283433 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGAACACGGCTGCTGGAATTCCA[A/G]AACCTAAGGGCTCGTGGGAGCTGTC
- Phenotype
-
MIM: 606514
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYTH4
PubMed Links
Gene Details
- Gene
- CYTH4
- Gene Name
- cytohesin 4
View Full Product Details