Product Details
- SNP ID
-
hCV30063766
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:36320824 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTGCACCCTCTCTGCCGCCTAA[A/G]GCCACATCTCTCTAACTCTTTACCT
- Phenotype
-
MIM: 616191
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLGAP4
PubMed Links
Gene Details
- Gene
- DLGAP4
- Gene Name
- DLG associated protein 4
View Full Product Details