Product Details
- SNP ID
-
rs10426308
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53385577 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCACATGCCTGCCGTCTCAGTTA[G/T]TGTGAAGTCTGTGCAGGGAGGATGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF525
PubMed Links
Gene Details
- Gene
- ZNF525
- Gene Name
- zinc finger protein 525
There are no transcripts associated with this gene.
- Gene
- ZNF765
- Gene Name
- zinc finger protein 765
There are no transcripts associated with this gene.
View Full Product Details