Product Details

SNP ID
rs6027204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59933085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGCAGCGCCAACCCCCCTTCAG[A/G]TGTCCCGGAGACAGGCGAGCAGCGG
Phenotype
MIM: 603326 MIM: 604105
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM217B PubMed Links

Gene Details

Gene
FAM217B
Gene Name
family with sequence similarity 217 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190826.1 Intron NP_001177755.1
NM_001190827.1 Intron NP_001177756.1
NM_022106.2 Intron NP_071389.1
XM_011528985.2 Intron XP_011527287.1
XM_011528986.2 Intron XP_011527288.1
Gene
PPP1R3D
Gene Name
protein phosphatase 1 regulatory subunit 3D
There are no transcripts associated with this gene.

Gene
SYCP2
Gene Name
synaptonemal complex protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014258.3 Intron NP_055073.2
XM_011528487.2 Intron XP_011526789.1
XM_011528488.2 Intron XP_011526790.1
XM_011528489.2 Intron XP_011526791.1
XM_011528490.2 Intron XP_011526792.1
XM_011528493.2 Intron XP_011526795.1
XM_017027586.1 Intron XP_016883075.1
XM_017027587.1 Intron XP_016883076.1
XM_017027588.1 Intron XP_016883077.1
XM_017027589.1 Intron XP_016883078.1
XM_017027590.1 Intron XP_016883079.1
XM_017027591.1 Intron XP_016883080.1
XM_017027592.1 Intron XP_016883081.1

View Full Product Details