Product Details

SNP ID
rs10496181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71070267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGTTTCGAGGACTAGGTGAAGC[A/G]TAAGTAGAGTCTTGCAGGATAAGGA
Phenotype
MIM: 606828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAGK PubMed Links

Gene Details

Gene
NAGK
Gene Name
N-acetylglucosamine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017567.4 Intron NP_060037.3
XM_005264412.4 Intron XP_005264469.1
XM_005264413.1 Intron XP_005264470.1
XM_005264415.2 Intron XP_005264472.1
XM_011532957.1 Intron XP_011531259.1
XM_017004440.1 Intron XP_016859929.1

View Full Product Details