Product Details

SNP ID
rs7010621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:57999374 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTTTTAAAAATACAATTCACAG[A/G]CATATGGGAAATTTTTGAGTGGTTG
Phenotype
MIM: 611394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM110B PubMed Links

Gene Details

Gene
FAM110B
Gene Name
family with sequence similarity 110 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147189.2 Intron NP_671722.1
XM_005251324.2 Intron XP_005251381.1
XM_005251325.3 Intron XP_005251382.1
XM_005251326.2 Intron XP_005251383.1
XM_017013948.1 Intron XP_016869437.1

View Full Product Details