Product Details

SNP ID
rs7225128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58559493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGAACACCCTCTGATTTCCTTCT[C/T]TCTGGTGTAATGCTATCTTCCCTTT
Phenotype
MIM: 605792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEPT4-AS1 PubMed Links

Gene Details

Gene
SEPT4-AS1
Gene Name
SEPT4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX14
Gene Name
testis expressed 14, intercellular bridge forming factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201457.1 4567 Silent Mutation GAA,GAG E1455E NP_001188386.1
NM_031272.4 4567 Silent Mutation GAA,GAG E1409E NP_112562.3
NM_198393.3 4567 Silent Mutation GAA,GAG E1449E NP_938207.2
XM_011525029.2 4567 Silent Mutation GAA,GAG E1490E XP_011523331.1
XM_017024860.1 4567 Silent Mutation GAA,GAG E1268E XP_016880349.1
XM_017024861.1 4567 Silent Mutation GAA,GAG E1057E XP_016880350.1

View Full Product Details