Product Details

SNP ID
rs9381498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47000139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAATCAGAAAACCCGATCGAATA[C/T]TGAGCATAATTTCTTCATTGACATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRF1 PubMed Links

Gene Details

Gene
ADGRF1
Gene Name
adhesion G protein-coupled receptor F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025048.3 2688 Intron NP_079324.2
NM_153840.3 2688 UTR 3 NP_722582.2
XM_011514469.1 2688 UTR 3 XP_011512771.1
XM_017010735.1 2688 UTR 3 XP_016866224.1
XM_017010736.1 2688 UTR 3 XP_016866225.1

View Full Product Details