Product Details

SNP ID
rs9381028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41057342 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAAGAGAAGGCAGCGCACTGA[A/C]TCTGGCCAACCTGCCTTCCACAGAG
Phenotype
MIM: 604797 MIM: 614393
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC2 PubMed Links

Gene Details

Gene
APOBEC2
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006789.3 Intron NP_006780.1
Gene
OARD1
Gene Name
O-acyl-ADP-ribose deacylase 1
There are no transcripts associated with this gene.

View Full Product Details